4-Bromo-3-chlorophenylacetylene 95%

Leverantör: Apollo Scientific

OR400945-1G OR400945-250MG
APOSOR400945-1GEA 7175 SEK
APOSOR400945-1G APOSOR400945-250MG
4-Bromo-3-chlorophenylacetylene 95%
4-Bromo-3-chlorophenylacetylene
Formel: C₈H₄BrCl
MW: 215.474 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD30086050
CAS nummer: 313511-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR