4-Bromo-3-methoxybenzenesulphonyl chloride 95%
Leverantör: Apollo Scientific
Fara
Synonyms: 4-Bromo-3-methoxybenzene-1-sulphonyl chloride
OR400875-1G OR400875-250MG OR400875-5G
APOSOR400875-1GEA 565 SEK
APOSOR400875-1G APOSOR400875-250MG APOSOR400875-5G
4-Bromo-3-methoxybenzenesulphonyl chloride 95%
4-Bromo-3-methoxybenzenesulphonyl chloride
Formel: C₇H₆BrClO₃S
Storage Temperature: Omgivande
MDL Number: MFCD14702923
CAS nummer: 1215295-91-2
UN: 1759
ADR: 8,III