Methyl 4-bromo-2-formylbenzoate
Leverantör: Apollo Scientific
|
Varning
|
OR400781-1G
OR400781-250MG
OR400781-5G
APOSOR400781-1GEA
2975
SEK
APOSOR400781-1G
APOSOR400781-250MG
APOSOR400781-5G
Methyl 4-bromo-2-formylbenzoate
Methyl 4-bromo-2-formylbenzoate
Formel:
C₉H₇BrO₃ MW: 243.054 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD16036574 CAS nummer: 1260795-42-3 |