2-Bromo-6-formylbenzoic acid 95%
Leverantör: Apollo Scientific
Varning
OR400774-1G OR400774-250MG
APOSOR400774-1GEA 8475 SEK
APOSOR400774-1G APOSOR400774-250MG
2-Bromo-6-formylbenzoic acid 95%
2-Bromo-6-formylbenzoic acid
Formel: C₈H₅BrO₃
MW: 229.027 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28357172
CAS nummer: 1245915-98-3