Methyl 3-bromo-4-formylbenzoate
Leverantör: Apollo Scientific
Varning
OR400764-1G OR400764-250MG
APOSOR400764-1GEA 2200 SEK
APOSOR400764-1G APOSOR400764-250MG
Methyl 3-bromo-4-formylbenzoate
Methyl 3-bromo-4-formylbenzoate
Formel: C₉H₇BrO₃
MW: 243.054 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16036864
CAS nummer: 90484-53-0