3-Bromo-4-formylbenzoic acid 97%
Leverantör: Apollo Scientific
OR400758-1G OR400758-5G
APOSOR400758-1GEA 960 SEK
APOSOR400758-1G APOSOR400758-5G
3-Bromo-4-formylbenzoic acid 97%
3-Bromo-4-formylbenzoic acid
Formel: C₈H₅BrO₃
MW: 229,027 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23135693
CAS nummer: 91760-66-6