3-Bromo-4-nitrobenzenesulphonyl chloride

Leverantör: Apollo Scientific
Fara

OR400681-1G OR400681-250MG OR400681-5G
APOSOR400681-1GEA 5475 SEK
APOSOR400681-1G APOSOR400681-250MG APOSOR400681-5G
3-Bromo-4-nitrobenzenesulphonyl chloride
3-Bromo-4-nitrobenzenesulphonyl chloride
Formel: C₆H₃BrClNO₄S
Storage Temperature: Omgivande
MDL Number: MFCD12197192
CAS nummer: 1181458-26-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR