2-Amino-5-bromo-3-chlorobenzoic acid
Leverantör: Apollo Scientific
Varning
OR400652-1G OR400652-250MG
APOSOR400652-1GEA 1850 SEK
APOSOR400652-1G APOSOR400652-250MG
2-Amino-5-bromo-3-chlorobenzoic acid
2-Amino-5-bromo-3-chlorobenzoic acid
Formel: C₇H₅BrClNO₂
Storage Temperature: Omgivande
MDL Number: MFCD00839522
CAS nummer: 58026-21-4