4-Bromo-5-methoxy-2-nitroaniline 98%
Leverantör: Apollo Scientific
Varning
OR400540-1G OR400540-5G
APOSOR400540-1GEA 1550 SEK
APOSOR400540-1G APOSOR400540-5G
4-Bromo-5-methoxy-2-nitroaniline 98%
4-Bromo-5-methoxy-2-nitroaniline
Formel: C₇H₇BrN₂O₃
MW: 247,046 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD13659425
CAS nummer: 173312-36-2