2-Bromo-3-iodobenzyl bromide
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Bromo-1-(bromomethyl)-3-iodobenzene
OR400517-5G OR400517-1G OR400517-250MG
APOSOR400517-5GEA 5000 SEK
APOSOR400517-5G APOSOR400517-1G APOSOR400517-250MG
2-Bromo-3-iodobenzyl bromide
2-Bromo-3-iodobenzyl bromide
Formel: C₇H₅Br₂I
MW: 375.827 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12025461
CAS nummer: 1261649-03-9
UN: 3261
ADR: 8 III