3-Vinylthiophene
Leverantör: Apollo Scientific
|
Varning
|
OR400379-1G
OR400379-5G
APOSOR400379-1GEA
2400
SEK
APOSOR400379-1G
APOSOR400379-5G
3-Vinylthiophene
3-Vinylthiophene
Formel:
C₆H₆S Storage Temperature: Kylskåp |
MDL Number:
MFCD07438049 CAS nummer: 13679-64-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...