4-Bromo-2-chloro-6-nitrophenol
Leverantör: Apollo Scientific
Varning
OR400331-1G OR400331-25G OR400331-5G
APOSOR400331-1GEA 260 SEK
APOSOR400331-1G APOSOR400331-25G APOSOR400331-5G
4-Bromo-2-chloro-6-nitrophenol
4-Bromo-2-chloro-6-nitrophenol
Formel: C₆H₃BrClNO₃
Storage Temperature: Omgivande
MDL Number: MFCD09835006
CAS nummer: 58349-01-2