2-Bromo-4-(tert-octyl)phenol
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Bromo-4-(1,1,3,3-tetramethylbut-1-yl)phenol, 2-Bromo-4-(2,4,4-trimethylpent-2-yl)phenol, 2-Bromo-4-(2,4,4-trimethylpentan-2-yl)phenol
OR40033-25G OR40033-5G OR40033-100G
APOSOR40033-25GEA 360 SEK
APOSOR40033-25G APOSOR40033-5G APOSOR40033-100G
2-Bromo-4-(tert-octyl)phenol
2-Bromo-4-(tert-octyl)phenol
Formel: C₁₄H₂₁BrO
MW: 285,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09907053
CAS nummer: 57835-35-5