2-Bromo-5-chloro-3-iodobenzoic acid
Leverantör: Apollo Scientific
Varning
OR400315-1G OR400315-250MG
APOSOR400315-1GEA 6600 SEK
APOSOR400315-1G APOSOR400315-250MG
2-Bromo-5-chloro-3-iodobenzoic acid
2-Bromo-5-chloro-3-iodobenzoic acid
Formel: C₇H₃BrClIO₂
MW: 361.359 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07780677
CAS nummer: 501009-09-2