7-Bromo-4-chloro-1,3-benzoxazole
Leverantör: Apollo Scientific
Varning
OR400238-1G OR400238-250MG
APOSOR400238-1GEA 7200 SEK
APOSOR400238-1G APOSOR400238-250MG
7-Bromo-4-chloro-1,3-benzoxazole
7-Bromo-4-chloro-1,3-benzoxazole
Formel: C₇H₃BrClNO
MW: 232,462 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28040522
CAS nummer: 1820707-32-1