Methyl 2-bromo-6-iodobenzoate 95%
Leverantör: Apollo Scientific
Fara
OR400123-10G OR400123-1G OR400123-5G
APOSOR400123-10GEA 5450 SEK
APOSOR400123-10G APOSOR400123-1G APOSOR400123-5G
Methyl 2-bromo-6-iodobenzoate 95%
Methyl 2-bromo-6-iodobenzoate
Formel: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11870176
CAS nummer: 1261840-81-6
UN: 2811
ADR: 6.1 III