2,4-Dinitrobenzaldoxime
Leverantör: Apollo Scientific
Varning
OR3936-1G OR3936-5G
APOSOR3936-1GEA 310 SEK
APOSOR3936-1G APOSOR3936-5G
2,4-Dinitrobenzaldoxime
2,4-Dinitrobenzaldoxime
Formel: C₇H₅N₃O₅
Storage Temperature: Omgivande
MDL Number: MFCD01070627
CAS nummer: 3236-33-7