1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Carboxy-1-methyl-3-phenyl-1H-pyrazole
OR3884-1G OR3884-5G OR3884-10G
APOSOR3884-1GEA 1600 SEK
APOSOR3884-1G APOSOR3884-5G APOSOR3884-10G
1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid 98%
1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid
Formel: C₁₁H₁₀N₂O₂
MW: 202,209 g/mol
Smältpunkt: 185,5…187 °C
Storage Temperature: Omgivande
MDL Number: MFCD08271939
CAS nummer: 10250-64-3