(Z)-Isopropyl 3-iodoacrylate ≥98%

Leverantör: Apollo Scientific

Synonyms: Isopropyl (Z)-3-iodoacrylate

OR370092-1G OR370092-5G
APOSOR370092-1GEA 5825 SEK
APOSOR370092-1G APOSOR370092-5G
(Z)-Isopropyl 3-iodoacrylate ≥98%
(Z)-Isopropyl 3-iodoacrylate
Formel: C₆H₉IO₂
MW: 240,039 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD30537663
CAS nummer: 1333154-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR