N,2-Dimethylbutan-2-amine ≥98%
Leverantör: Apollo Scientific
OR361733-1G OR361733-250MG
APOSOR361733-1GEA 10925 SEK
APOSOR361733-1G APOSOR361733-250MG
N,2-Dimethylbutan-2-amine ≥98%
N,2-Dimethylbutan-2-amine
Formel: C₆H₁₅N
MW: 101.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00135017
CAS nummer: 2978-64-5