3-Methoxy-1-propyn-1-ylboronic acid, pinacol ester 96%
Leverantör: Apollo Scientific
OR361396-1G OR361396-5G
APOSOR361396-1GEA 1575 SEK
APOSOR361396-1G APOSOR361396-5G
3-Methoxy-1-propyn-1-ylboronic acid, pinacol ester 96%
3-Methoxy-1-propyn-1-ylboronic acid, pinacol ester
Formel: C₁₀H₁₇BO₃
MW: 196,051 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08705273
CAS nummer: 634196-63-7