4-(Methoxycarbonyl)-2-methylphenylboronic acid 98%
Leverantör: Apollo Scientific
Synonyms: (4-(Methoxycarbonyl)-2-methylphenyl)boronic acid
OR360222-5G OR360222-1G OR360222-250MG
APOSOR360222-5GEA 4925 SEK
APOSOR360222-5G APOSOR360222-1G APOSOR360222-250MG
4-(Methoxycarbonyl)-2-methylphenylboronic acid 98%
4-(Methoxycarbonyl)-2-methylphenylboronic acid
Formel: C₉H₁₁BO₄
MW: 193,992 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16036117
CAS nummer: 158429-38-0