1H-1,2,3-Benzotriazol-5-ylboronic acid 95%
Leverantör: Apollo Scientific
Varning
OR360093-1G OR360093-250MG
APOSOR360093-1GEA 11325 SEK
APOSOR360093-1G APOSOR360093-250MG
1H-1,2,3-Benzotriazol-5-ylboronic acid 95%
1H-1,2,3-Benzotriazol-5-ylboronic acid
Formel: C₆H₆BN₃O₂
MW: 162,942 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD10697877
CAS nummer: 183282-45-3