2-Methoxy-5-(methylsulphonyl)benzoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Carboxy-4-(methylsulphonyl)anisole, 5-(Methylsulphonyl)-o-anisic acid, 3-Carboxy-4-methoxyphenyl methyl sulphone
OR3569-25G OR3569-5G OR3569-100G
APOSOR3569-25GEA 565 SEK
APOSOR3569-25G APOSOR3569-5G APOSOR3569-100G
2-Methoxy-5-(methylsulphonyl)benzoic acid
2-Methoxy-5-(methylsulphonyl)benzoic acid
Formel: C₉H₁₀O₅S
MW: 230.238 g/mol
Smältpunkt: 190…193 °C
Storage Temperature: Omgivande
MDL Number: MFCD01317544
CAS nummer: 50390-76-6