1-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
Leverantör: Apollo Scientific
Synonyms: 1-Isopropyl-1H-pyrazole-5-boronic acid pinacol ester
OR350205-1G OR350205-5G
APOSOR350205-1GEA 890 SEK
APOSOR350205-1G APOSOR350205-5G
1-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel: C₁₂H₂₁BN₂O₂
MW: 236.118 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17167306
CAS nummer: 1282518-60-8