1-Aminopyrrolidin-2-one hydrochloride
Leverantör: Apollo Scientific
OR350009-1G OR350009-250MG OR350009-5G
APOSOR350009-1GEA 2125 SEK
APOSOR350009-1G APOSOR350009-250MG APOSOR350009-5G
1-Aminopyrrolidin-2-one hydrochloride
1-Aminopyrrolidin-2-one hydrochloride
Formel: C₄H₈N₂O·ClH
MW: 136.58 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09258892
CAS nummer: 20386-22-5