4-Bromopyridine 1-oxide 95+%
Leverantör: Apollo Scientific
OR346541-1G
OR346541-5G
OR346541-250MG
APOSOR346541-1GEA
2425
SEK
APOSOR346541-1G
APOSOR346541-5G
APOSOR346541-250MG
4-Bromopyridine 1-oxide 95+%
4-Bromopyridine 1-oxide
Formel:
C₅H₄BrNO MW: 173,995 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00234039 CAS nummer: 14248-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...