2-(3,4-Dichlorophenyl)-5-methyl-[1,3,4]oxadiazole 95+%
Leverantör: Apollo Scientific
OR346442-1G OR346442-5G
APOSOR346442-1GEA 2150 SEK
APOSOR346442-1G APOSOR346442-5G
2-(3,4-Dichlorophenyl)-5-methyl-[1,3,4]oxadiazole 95+%
2-(3,4-Dichlorophenyl)-5-methyl-[1,3,4]oxadiazole
Formel: C₉H₆Cl₂N₂O
MW: 229,063 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19981450
CAS nummer: 41420-95-5