2-Chloropyridine-3,5-dicarboxaldehyde 95+%
Leverantör: Apollo Scientific
OR346412-1G OR346412-500MG OR346412-5G
APOSOR346412-1GEA 3125 SEK
APOSOR346412-1G APOSOR346412-500MG APOSOR346412-5G
2-Chloropyridine-3,5-dicarboxaldehyde 95+%
2-Chloropyridine-3,5-dicarboxaldehyde
Formel: C₇H₄ClNO₂
MW: 169,565 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12131128
CAS nummer: 129216-07-5