1,2,5-Trimethyl-pyrrolidine-2,5-dicarbonitrile 95+%

Leverantör: Apollo Scientific

OR346402-1G OR346402-5G
APOSOR346402-1GEA 1825 SEK
APOSOR346402-1G APOSOR346402-5G
1,2,5-Trimethyl-pyrrolidine-2,5-dicarbonitrile 95+%
1,2,5-Trimethyl-pyrrolidine-2,5-dicarbonitrile
Formel: C₉H₁₃N₃
MW: 163,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11845737
CAS nummer: 57013-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR