1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde 95+%
Leverantör: Apollo Scientific
OR346399-1G
OR346399-500MG
APOSOR346399-1GEA
3750
SEK
APOSOR346399-1G
APOSOR346399-500MG
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde 95+%
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde
Formel:
C₁₂H₁₁BrN₂O MW: 279,133 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD11845716 CAS nummer: 294877-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...