3-(5-tert-Butyl-isoxazol-3-yl)-3-oxo-propionitrile
Leverantör: Apollo Scientific
OR346366-1G
OR346366-5G
APOSOR346366-1GEA
0
SEK
APOSOR346366-1G
APOSOR346366-5G
3-(5-tert-Butyl-isoxazol-3-yl)-3-oxo-propionitrile
3-(5-tert-Butyl-isoxazol-3-yl)-3-oxo-propionitrile
Formel:
C₁₀H₁₂N₂O₂ MW: 192,214 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD24386152 CAS nummer: 1423915-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...