4-Bromo-3,5-dimethyl-1-phenyl-1H-pyrazole
Leverantör: Apollo Scientific
OR346290-1G OR346290-5G
APOSOR346290-1GEA 0 SEK
APOSOR346290-1G APOSOR346290-5G
4-Bromo-3,5-dimethyl-1-phenyl-1H-pyrazole
4-Bromo-3,5-dimethyl-1-phenyl-1H-pyrazole
Formel: C₁₁H₁₁BrN₂
MW: 251,122 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00462174
CAS nummer: 51294-75-8