2-Chloromethyl-[1,3]dioxane
Leverantör: Apollo Scientific
OR346179-1G OR346179-500MG
APOSOR346179-1GEA 0 SEK
APOSOR346179-1G APOSOR346179-500MG
2-Chloromethyl-[1,3]dioxane
2-Chloromethyl-[1,3]dioxane
Formel: C₅H₉ClO₂
MW: 136,577 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00154081
CAS nummer: 5695-72-7