8-Chloro-11-[(n-methyl)-4-piperidylidene]-6,11-dihydro-5h-benzo [5,6] cyclohepta[1,2b]pyridine 97%
Leverantör: Apollo Scientific
Synonyms: 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
OR345568-1G OR345568-5G OR345568-25G
APOSOR345568-1GEA 155 SEK
APOSOR345568-1G APOSOR345568-5G APOSOR345568-25G
8-Chloro-11-[(n-methyl)-4-piperidylidene]-6,11-dihydro-5h-benzo [5,6] cyclohepta[1,2b]pyridine 97%
8-Chloro-11-[(n-methyl)-4-piperidylidene]-6,11-dihydro-5h-benzo [5,6] cyclohepta[1,2b]pyridine
Formel: C₂₀H₂₁ClN₂
MW: 324,847 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06411084
CAS nummer: 38092-89-6