2-Phenylbenzofuran
Leverantör: Apollo Scientific
|
Varning
|
OR340156-1G
OR340156-250MG
APOSOR340156-1GEA
6675
SEK
APOSOR340156-1G
APOSOR340156-250MG
2-Phenylbenzofuran
2-Phenylbenzofuran
Formel:
C₁₄H₁₀O MW: 194.229 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00187273 CAS nummer: 1839-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...