4H,5H-Pyrazolo[1,5-a]pyrimidin-5-one 97%
Leverantör: Apollo Scientific
OR33560-1G OR33560-5G
APOSOR33560-1GEA 995 SEK
APOSOR33560-1G APOSOR33560-5G
4H,5H-Pyrazolo[1,5-a]pyrimidin-5-one 97%
4H,5H-Pyrazolo[1,5-a]pyrimidin-5-one
Formel: C₆H₅N₃O
MW: 135,123 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14581682
CAS nummer: 29274-22-4