Phenyl N-(4-acetylphenyl)carbamate 95%

Leverantör: Apollo Scientific

OR33156-1G OR33156-5G
APOSOR33156-1GEA 2425 SEK
APOSOR33156-1G APOSOR33156-5G
Phenyl N-(4-acetylphenyl)carbamate 95%
Phenyl N-(4-acetylphenyl)carbamate
Formel: C₁₅H₁₃NO₃
MW: 255,269 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00532616
CAS nummer: 99134-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR