5-Oxo-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylic acid 95%
Leverantör: Apollo Scientific
OR32581-1G
OR32581-5G
APOSOR32581-1GEA
2425
SEK
APOSOR32581-1G
APOSOR32581-5G
5-Oxo-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylic acid 95%
5-Oxo-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylic acid
Formel:
C₁₃H₁₅NO₃ MW: 233,263 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD08690908 CAS nummer: 915302-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...