1-(5-Bromo-7-methoxy-1-benzofuran-2-yl)ethan-1-one 95%

Leverantör: Apollo Scientific

OR32454-1G OR32454-5G
APOSOR32454-1GEA 2575 SEK
APOSOR32454-1G APOSOR32454-5G
1-(5-Bromo-7-methoxy-1-benzofuran-2-yl)ethan-1-one 95%
1-(5-Bromo-7-methoxy-1-benzofuran-2-yl)ethan-1-one
Formel: C₁₁H₉BrO₃
MW: 269,091 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02102416
CAS nummer: 150612-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR