(2E)-3-(Dimethylamino)-1-(4-methylphenyl)prop-2-en-1-one 95%
Leverantör: Apollo Scientific
OR32178-1G OR32178-5G
APOSOR32178-1GEA 3875 SEK
APOSOR32178-1G APOSOR32178-5G
(2E)-3-(Dimethylamino)-1-(4-methylphenyl)prop-2-en-1-one 95%
(2E)-3-(Dimethylamino)-1-(4-methylphenyl)prop-2-en-1-one
Formel: C₁₂H₁₅NO
MW: 189,254 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00121190
CAS nummer: 502841-03-4