(2-(Piperidin-4-yl)phenyl)methanol hydrochloride
Leverantör: Apollo Scientific
OR321489-1G OR321489-250MG
APOSOR321489-1GEA 7700 SEK
APOSOR321489-1G APOSOR321489-250MG
(2-(Piperidin-4-yl)phenyl)methanol hydrochloride
(2-(Piperidin-4-yl)phenyl)methanol hydrochloride
Formel: C₁₂H₁₇NO·ClH
MW: 227.73 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD29075591
CAS nummer: 371981-27-0