Ethyl 1H-pyrrolo[3,2-b]pyridine-2-carboxylate 97%
Leverantör: Apollo Scientific
OR321483-1G OR321483-5G
APOSOR321483-1GEA 945 SEK
APOSOR321483-1G APOSOR321483-5G
Ethyl 1H-pyrrolo[3,2-b]pyridine-2-carboxylate 97%
Ethyl 1H-pyrrolo[3,2-b]pyridine-2-carboxylate
Formel: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11053752
CAS nummer: 17288-32-3