Methyl (R)-4-(piperidin-2-yl)benzoate hydrochloride 97%
Leverantör: Apollo Scientific
Synonyms: (R)-Methyl 4-(piperidin-2-yl)benzoate hydrochloride
OR321383-100MG OR321383-1G
APOSOR321383-100MGEA 1725 SEK
APOSOR321383-100MG APOSOR321383-1G
Methyl (R)-4-(piperidin-2-yl)benzoate hydrochloride 97%
Methyl (R)-4-(piperidin-2-yl)benzoate hydrochloride
Formel: C₁₃H₁₇NO₂·ClH
MW: 255.741 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12910877
CAS nummer: 1391574-76-7