4-(4-Bromo-1H-pyrazol-1-yl)piperidine hydrochloride 97%
Leverantör: Apollo Scientific
Varning
OR321335-100MG OR321335-250MG
APOSOR321335-100MGEA 1200 SEK
APOSOR321335-100MG APOSOR321335-250MG
4-(4-Bromo-1H-pyrazol-1-yl)piperidine hydrochloride 97%
4-(4-Bromo-1H-pyrazol-1-yl)piperidine hydrochloride
Formel: C₈H₁₂BrN₃·ClH
MW: 266.566 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18432580
CAS nummer: 1263378-44-4