4-(2-Ethylphenyl)piperidine hydrochloride > 95%
Leverantör: Apollo Scientific
Varning
OR321245-100MG OR321245-20MG OR321245-50MG
APOSOR321245-100MGEA 5075 SEK
APOSOR321245-100MG APOSOR321245-20MG APOSOR321245-50MG
4-(2-Ethylphenyl)piperidine hydrochloride > 95%
4-(2-Ethylphenyl)piperidine hydrochloride
Formel: C₁₃H₁₉N·ClH
MW: 225,758 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD30099247
CAS nummer: 74150-51-9