tert-Butyl (R)-(piperidin-2-ylmethyl)carbamate 97%

Leverantör: Apollo Scientific
Varning

OR321208-1G OR321208-5G
APOSOR321208-1GEA 895 SEK
APOSOR321208-1G APOSOR321208-5G
tert-Butyl (R)-(piperidin-2-ylmethyl)carbamate 97%
tert-Butyl (R)-(piperidin-2-ylmethyl)carbamate
Formel: C₁₁H₂₂N₂O₂
MW: 214,305 g/mol
Smältpunkt: 109…114 °C
Storage Temperature: Kylskåp
MDL Number: MFCD06738954
CAS nummer: 139004-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR