Naphtho[2,3-d][1,3]dioxole-6-carbonitrile 95+%
Leverantör: Apollo Scientific
OR318030-1G OR318030-10G OR318030-5G
APOSOR318030-1GEA 5025 SEK
APOSOR318030-1G APOSOR318030-10G APOSOR318030-5G
Naphtho[2,3-d][1,3]dioxole-6-carbonitrile 95+%
Naphtho[2,3-d][1,3]dioxole-6-carbonitrile
Formel: C₁₂H₇NO₂
MW: 197,189 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11226764
CAS nummer: 107401-98-9