3-Methyl-1H-pyrazole-5-carboxamide 95+%
Leverantör: Apollo Scientific
|
Varning
|
OR318021-1G
OR318021-10G
OR318021-5G
APOSOR318021-1GEA
2500
SEK
APOSOR318021-1G
APOSOR318021-10G
APOSOR318021-5G
3-Methyl-1H-pyrazole-5-carboxamide 95+%
3-Methyl-1H-pyrazole-5-carboxamide
Formel:
C₅H₇N₃O MW: 125,129 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD06808782 CAS nummer: 4027-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...