2-Azaspiro[3.5]nonan-7-one, NBOC protected 95+%
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 7-oxo-2-azaspiro[3.5]nonane-2-carboxylate
OR317318-1G OR317318-500MG
APOSOR317318-1GEA 9000 SEK
APOSOR317318-1G APOSOR317318-500MG
2-Azaspiro[3.5]nonan-7-one, NBOC protected 95+%
2-Azaspiro[3.5]nonan-7-one, NBOC protected
Formel: C₁₃H₂₁NO₃
MW: 239,311 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22566155
CAS nummer: 1363381-22-9